missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Standard, 0.5% w/w Recommended Storage: 2°C to 8°C (36°F to 46°F)., Reagents

Catalog No. 01335599
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335599 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335599 Supplier Labchem Inc Supplier No. CC2044001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Citric Acid Standard, 0.5% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Citric Acid Standard, 0.5% w/w
pH 2.6
CAS 7732-18-5, 77-92-9
Boiling Point 100°C
Color Colorless
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.001
Molecular Formula C6H8O7/H2O
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
Percent Purity 0.5%
Grade Reagent
Show More Show Less