missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, Anhydrous, Granular, FCC, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18601713
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601713 500 g Poly Bottle
18601714 2.5 kg Poly Pail
18601715 12 kg Poly Pail
18601716 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601713 Supplier Spectrum Chemical Mfg Cor Supplier No. C1281500GM
Only null left

C1281, 77-92-9, C6H8O7

Spectrum™ Chemical Citric Acid, Anhydrous, Granular, FCC is the most widely used organic acid and pH-control agent in foods and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

CAS Max % 1
Percent Purity 99.5 to 100.5%
Grade FCC
Quantity 500 g
Ignition Residue 0.0005
Packaging Poly Bottle