missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Anhydrous, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| Color | White |
| CAS Max % | 100.0 |
| Quantity | 500 g |
| Packaging | White Poly Wide Mouth Bottle |
| Physical Form | Granules |
| Percent Purity | Min. 99.5% |
| Grade | ACS |
| Chemical Name or Material | Citric Acid Anhydrous |
Safety and Handling
| ShelfLife | 60 Months |