missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, Anhydrous, 99.5 to 100.5% (Dry Basis)

Catalog No. AAJ13729A1
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ13729A1 1 kg Plastic bottle
AAJ1372936 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ13729A1 Supplier Thermo Fisher Scientific Supplier No. J13729A1
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77-92-9 | C6H8O7 | 192.123 g/mol

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.123
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Melting Point 153°C to 154°C
Density 1.665
Quantity 1 kg
Packaging Plastic bottle
Physical Form Crystalline Powder
Specific Gravity 1.665
Beilstein 782061
Formula Weight 192.12
Chemical Name or Material Citric Acid Anhydrous
Hazard Category H319-H335-H500
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 201-069-1
RTECSNumber GE7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only