missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, 15% (w/w), Reagents

Catalog No. 01335596
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335596 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335596 Supplier Labchem Inc Supplier No. CC204700250C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Citric Acid, 15% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 77-92-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym 2-hydroxypropane-1,2,3-tricarboxylic Acid
IUPAC Name water
SMILES O

Specifications

pH <2
Boiling Point 100°C
Color Colorless
Quantity 250 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.0645
Percent Purity 15%
Grade Reagent
Chemical Name or Material Citric Acid, 15% (w/w)