missing translation for 'onlineSavingsMsg'
Learn More

Citral (cis- and trans- mixture) 96.0+%, TCI America™

Catalog No. D076225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D076225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D076225ML Supplier TCI America Supplier No. D076225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5392-40-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00006997
InChI Key WTEVQBCEXWBHNA-JXMROGBWSA-N
Synonym citral, geranial, trans-citral, 3,7-dimethylocta-2,6-dienal, geranialdehyde, e-citral, citral a, 2e-3,7-dimethylocta-2,6-dienal, alpha-citral, lemsyn gb
PubChem CID 638011
ChEBI CHEBI:16980
IUPAC Name (2E)-3,7-dimethylocta-2,6-dienal
SMILES CC(C)=CCC\C(C)=C\C=O

Specifications

Color Yellow
Boiling Point 229°C
Quantity 25 mL
Formula Weight 152.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Citral (cis- and trans- mixture)
EINECSNumber (2)-0515
RTECSNumber RG5075000
TSCA Yes