missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citral, cis + trans, 95%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Melting Point | <-10°C |
| Density | 0.888 |
| Boiling Point | 228°C to 229°C |
| Flash Point | 98°C (208°F) |
| Refractive Index | 1.488 |
| Quantity | 50 g |
| Beilstein | 1721871 |
| Merck Index | 14,2322 |
| Formula Weight | 152.24 |
| Percent Purity | 95% |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H315-H317-H319 |
| Hazard Statement | GHS H Statement H315-H317 Causes skin irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c |
| EINECSNumber | 226-394-6 |
| RTECSNumber | RG5075000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only