Learn More
Citraconic anhydride, 98%
Description
Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Citraconic anhydride is used as a monomer for specialty unsaturated polyester resins. It is used to prepare bicyclic pyrrolidines, maleimides and co- and terpolymers. It is also used for the protection of N-terminal amino acids. Further, it is involved in the preparation of clothing ping acid and its amide. In addition to this, it is selectively used for the isolation of histidyl peptides.
Solubility
Miscible with water, alcohol, acetone and ether.
Notes
Moisture sensitive. Incompatible with bases, oxidizing agents and reducing agents.
Specifications
Specifications
| Melting Point | 7°C to 8°C |
| Density | 1.243 |
| Boiling Point | 213°C to 214°C |
| Flash Point | 101°C (213°F) |
| Refractive Index | 1.471 |
| Quantity | 500 g |
| UN Number | UN2810 |
| Beilstein | 1835 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Miscible with water,alcohol,acetone and ether. |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H311-H314-H317-H334 |
| missing translation for 'healthHazard2' | GHS H Statement H311-H315-H319 Toxic in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| missing translation for 'healthHazard3' | P260-P264b-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 210-459-0 |
| RTECSNumber | GE6825000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only