missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cisplatin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Cisplatin is an antineoplastic drug used for the treatment of cancer patients with a variety of different malignancies. Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 15663-27-1 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | Pt(NH3)2 Cl2 |
| MDL Number | MFCD00011623 |
| Melting Point | 270°C |
| Synonym | cis-Diammineplatinum(II) dichloride; cis-Dichlorodiammine platinum(II); cis-Platinum(II) diammine dichloride; Cisplatin |
| Type | Certified Reference Material |
| Quantity | 200 mg |
| Formula Weight | 300.05 |
| Grade | Certified Reference Material |