missing translation for 'onlineSavingsMsg'
Learn More

cis-6-Nonen-1-ol 90.0+%, TCI America™

Catalog No. N046910ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N046910ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N046910ML Supplier TCI America Supplier No. N046910ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35854-86-5
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00015388
InChI Key XJHRZBIBSSVCEL-ARJAWSKDSA-N
Synonym cis-6-nonen-1-ol, cis-6-nonenol, z-non-6-en-1-ol, z-6-nonen-1-ol, 6z-nonen-1-ol, 6z-non-6-en-1-ol, 6-nonen-1-ol, z, z-6-nonenol, 6z-6-nonen-1-ol
PubChem CID 5362792
IUPAC Name (Z)-non-6-en-1-ol
SMILES CCC=CCCCCCO

Specifications

Color Colorless
Boiling Point 112°C
Quantity 10 mL
Formula Weight 142.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-6-Nonen-1-ol
EINECSNumber (2)-0258
TSCA Yes