missing translation for 'onlineSavingsMsg'
Learn More

cis-4-tert-Butylcyclohexanol 98.0+%, TCI America™

Catalog No. B12705G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12705G 5 g
B12701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. B12705G missing translation for 'mfr' TCI America missing translation for 'supplierNo' B12705G

Chemical Identifiers

CAS 937-05-3
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00070476
InChI Key CCOQPGVQAWPUPE-UHFFFAOYSA-N
PubChem CID 7391
IUPAC Name 4-tert-butylcyclohexan-1-ol
SMILES CC(C)(C)C1CCC(CC1)O

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 5 g
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-tert-Butylcyclohexanol