missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Cyclohexene-1,2-dicarboxylic Acid 98.0+%, TCI America™

Catalog No. C0492500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0492500G 500 g
C049225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0492500G Supplier TCI America Supplier No. C0492500G

Chemical Identifiers

CAS 2305-26-2
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00070482
InChI Key ILUAAIDVFMVTAU-OLQVQODUSA-N
Synonym cis-4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 4-cyclohexene-1,2-dicarboxylic acid, cis, cis-delta4-tetrahydrophthalic acid, 4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel, rel-1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-rel-4-cyclohexene-1,2-dicarboxylic acid, cis-.delta.4-tetrahydrophthalic acid, cis-.delta.4-cyclohexene-1,2-dicarboxylic acid
PubChem CID 504243
IUPAC Name (1S,2R)-cyclohex-4-ene-1,2-dicarboxylic acid
SMILES C1C=CCC(C1C(=O)O)C(=O)O

Specifications

Melting Point 168°C
Color White
Quantity 500 g
Formula Weight 170.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-Cyclohexene-1,2-dicarboxylic Acid
EINECSNumber (3)-3316
TSCA Yes