missing translation for 'onlineSavingsMsg'
Learn More

cis-4,7,10,13,16,19-Docosahexaenoic Acid 97.0+%, TCI America™

Catalog No. D2226100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2226100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2226100MG Supplier TCI America Supplier No. D2226100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6217-54-5
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.496
MDL Number MFCD00065722
InChI Key MBMBGCFOFBJSGT-OBOJEMQYSA-N
Synonym cis-4,7,10,13,16,19-docosahexaenoic acid, 4,7,10,13,16,19-cis-docosahexaenoic acid, cis-4, 7, 10, 13, 16, 19-docosahexaenoic acid, 4z-docosa-4,7,10,13,16,19-hexaenoic acid
PubChem CID 57417355
IUPAC Name (4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
SMILES CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O

Specifications

Melting Point -45°C
Color Yellow
Quantity 100 mg
Formula Weight 328.50
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-4,7,10,13,16,19-Docosahexaenoic Acid
EINECSNumber (2)-0609
TSCA No
Recommended Storage Refrigerator