missing translation for 'onlineSavingsMsg'
Learn More

cis-4,7,10,13,16,19-Docosahexaenoic Acid, >99%, MP Biomedicals™

Catalog No. p-4405419
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15909725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15909725 Supplier MP Biomedicals Inc Supplier No. 0215909725

Chemical Identifiers

CAS 6217-54-5
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.496
MDL Number MFCD00065722
InChI Key MBMBGCFOFBJSGT-OBOJEMQYSA-N
PubChem CID 57417355
IUPAC Name (4E,7E,10E,13E,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
SMILES CCC=CCC=CCC=CCC=CCC=CCC=CCCC(=O)O

Specifications

Color Colorless
Assay Percent Range ≥98%
Quantity 25 mg
Synonym cis-4,7,10,13,16,19-docosahexaenoic acid, 4,7,10,13,16,19-cis-docosahexaenoic acid, cis-4, 7, 10, 13, 16, 19-docosahexaenoic acid, 4z-docosa-4,7,10,13,16,19-hexaenoic acid
Formula Weight 328.5
Physical Form Liquid
Chemical Name or Material cis-4,7,10,13,16,19-Docosahexaenoic Acid
Recommended Storage Store at - 0°C, protect from light, store under nitrogen