missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexen-1-yl Hexanoate 98.0+%, TCI America™

Numéro de catalogue. C091725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C091725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. C091725ML Fournisseur TCI America Code fournisseur C091725ML
Il en reste null

Identifiants chimiques

CAS 31501-11-8
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.31
MDL Number MFCD00036552,MFCD00036552
InChI Key RGACQXBDYBCJCY-ALCCZGGFSA-N
Synonym Hexanoic Acid cis-3-Hexen-1-yl Ester
PubChem CID 5352543
IUPAC Name (3Z)-hex-3-en-1-yl hexanoate
SMILES CCCCCC(=O)OCC\C=C/CC

Spécifications

Color Colorless
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-yl Hexanoate
EINECSNumber (2)-0760
RTECSNumber MO8380000
TSCA Yes