missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Pentene 95.0+%, TCI America™

Catalog No. P006510ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P006510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P006510ML Supplier TCI America Supplier No. P006510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627-20-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00063978
InChI Key QMMOXUPEWRXHJS-HYXAFXHYSA-N
Synonym cis-2-pentene, z-2-pentene, 2-pentene, z, cis-pentene, z-pent-2-ene, unii-54ur3xz4fc, cis-pentene-2, 54ur3xz4fc, 2-pentene, 2-pentene, 2z
PubChem CID 5326160
IUPAC Name (2Z)-pent-2-ene
SMILES CC\C=C/C

Specifications

Melting Point -180°C
Color Colorless
Boiling Point 37°C
Quantity 10 mL
UN Number 3295
Formula Weight 70.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2-Pentene
EINECSNumber (2)-0019
TSCA Yes