missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Hexene 95.0+%, TCI America™

Catalog No. H04045ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04045ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04045ML Supplier TCI America Supplier No. H04045ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7688-21-3
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00066520
InChI Key RYPKRALMXUUNKS-HYXAFXHYSA-N
Synonym cis-2-hexene, 2-hexene, z, z-2-hexene, z-hex-2-ene, 2-hexene, cis, unii-ui6o1h5j7j, 2-hexene-cis, 2z-hex-2-ene, ui6o1h5j7j, 2-hexene z
PubChem CID 643835
IUPAC Name (2Z)-hex-2-ene
SMILES CCC\C=C/C

Specifications

Color Yellow
Boiling Point 69°C
Quantity 5 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2-Hexene
EINECSNumber (2)-0031
TSCA No