missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-2-Aminocyclohexanol hydrochloride, 99+%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | cis-2-Aminocyclohexanol hydrochloride |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 99+% |
| Emballage | Glass bottle |
| Formule linéaire | H2NC6H10OH·HCl |
| Quantité | 5g |
| Renseignements sur la solubilité | Solubility in water: soluble. Other solubilities: soluble in methanol,hot ethanol |
| Poids de formule | 151.64 |
| Pourcentage de pureté | 99+% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only