Learn More
Graphic Controls Circular Charts
Graphic Controls Circular Charts, Diameter: 157.23mm, 6.19 in., Ink Writing Recorder Chart, Time Range: 7 Days, For Use With: For weksler, No. of Ranges: 1.0, Shape: Circular
Supplier: Graphic Controls 31405791
Description
Graphic Controls Circular Charts, Diameter: 157.23mm, 6.19 in., Ink Writing Recorder Chart, Time Range: 7 Days, For Use With: For weksler, No. of Ranges: 1.0, Shape: Circular
Identifiants chimiques
| Circular |
Spécifications
| Ink Writing Recorder Chart | |
| 7 in. | |
| 1 | |
| -100°C to +38°C |
| 177.8 mm | |
| Jewett RDR069 | |
| Circular | |
| 7 Days |