missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIRBP Mouse anti-Human, Clone:6E12B3, Abnova™
Mouse monoclonal antibody raised against recombinant human CIRBP.
Supplier: Abnova Corporation MAB17836
Spécifications
| CIRBP | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Flow Cytometry Immunocytochemistry Immunohistochemistry (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
| CIRBP | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6E12B3 | |
| cold inducible RNA binding protein | |
| In PBS (0.05% sodium azide) | |
| CIRP | |
| Recombinant protein corresponding to amino acids 1-90 of human CIRBP from E. coli. | |
| 1153 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |