missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Hydrochloride USP, Medisca™

$233.07 - $1070.33

Chemical Identifiers

CAS 86393-32-0
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
18-048-039
View Documents
Medisca Pharmaceutique Inc
053409
1 kg
Each for $1,070.33
Only null left
 
18-048-042
View Documents
Medisca Pharmaceutique Inc
053408
500 g
Each for $637.91
Only null left
 
18-048-103
View Documents
Medisca Pharmaceutique Inc
053405
100 g
Each for $233.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

86393-32-0

Specifications

α-Carboxybenzyl Penicillin Disodium Salt
86393-32-0
1 kg
USP
Crystals
Light Yellow to Yellow
98.0 to 102.0%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'