missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Hydrochloride USP, Medisca™
GREENER_CHOICE

Catalog No. 18048103
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048103 100 g
18048039 1 kg
18048042 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18048103 Supplier Medisca Pharmaceutique Inc Supplier No. 053405
Only null left

Chemical Identifiers

CAS 86393-32-0

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystals
Color Light Yellow to Yellow
Quantity 100 g
Percent Purity 98.0 to 102.0%
Grade USP