missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciprofibrate 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 118°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 289.15 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Ciprofibrate |
Sécurité et manipulation
| RTECSNumber | UF0880000 |
| TSCA | No |
| Recommended Storage | Refrigerator |