missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl bromide, predominantly trans, 95%

Catalog No. AAL0142006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0142006 5 g
AAL0142014 25 g
AAL0142022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0142006 Supplier Thermo Scientific Chemicals Supplier No. L0142006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4392-24-9 | C9H9Br | 197.075 g/mol

Cinnamyl bromide, is used in the synthesis of naftifine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl bromide, is used in the synthesis of naftifine.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 4392-24-9
Molecular Formula C9H9Br
Molecular Weight (g/mol) 197.075
MDL Number MFCD00000245
InChI Key RUROFEVDCUGKHD-QPJJXVBHSA-N
Synonym cinnamyl bromide, 3-bromoprop-1-en-1-yl benzene, e-3-bromoprop-1-en-1-yl benzene, 3-bromo-1-phenyl-1-propene, 3-bromo-1-phenyl propene-1, e-3-bromoprop-1-enyl benzene, 3-bromo-propenyl-benzene, e-cinnamyl bromide, trans-cinnamyl bromide
PubChem CID 5357478
IUPAC Name [(E)-3-bromoprop-1-enyl]benzene
SMILES C1=CC=C(C=C1)C=CCBr

Specifications

Melting Point 26°C to 29°C
Boiling Point 88°C to 90°C (1 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.621
Quantity 5 g
UN Number UN3261
Beilstein 605833
Sensitivity Light sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 197.08
Percent Purity 95%
Chemical Name or Material Cinnamyl bromide, predominantly trans
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 224-511-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only