missing translation for 'onlineSavingsMsg'
Learn More

Cilnidipine 98.0+%, TCI America™

Catalog No. C25645G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25645G 5 g
C25641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25645G Supplier TCI America Supplier No. C25645G

Chemical Identifiers

CAS 132203-70-4
Molecular Formula C27H28N2O7
Molecular Weight (g/mol) 492.528
MDL Number MFCD00865853
InChI Key KJEBULYHNRNJTE-DHZHZOJOSA-N
Synonym cilnidipine, cinalong, atelec, siscard, cilnidipine inn, cinaldipine, 3-cinnamyl 5-2-methoxyethyl 2,6-dimethyl-4-3-nitrophenyl-1,4-dihydropyridine-3,5-dicarboxylate, atelec tn, +--e-cinnamyl 2-methoxyethyl 1,4-dihydro-2,6-dimethyl-4-m-nitrophenyl-3,5-pyridinedicarboxylate, 1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 2-methoxyethyl 2e-3-phenyl-2-propenyl ester
PubChem CID 5282138
ChEBI CHEBI:31399
IUPAC Name 3-O-(2-methoxyethyl) 5-O-[(E)-3-phenylprop-2-enyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCC=CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OCCOC

Specifications

Melting Point 110°C
Color Yellow
Quantity 5 g
Formula Weight 492.53
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cilnidipine
RTECSNumber US7975550
TSCA No