missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cilnidipine, 99%

Supplier:  Thermo Scientific™ 461750050

Quantity 5g

 Afficher plus de versions de ce produit

Numéro de catalogue. AC461750050

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Identifiants chimiques

132203-70-4
492.52
5282138
C27H28N2O7
KJEBULYHNRNJTE-DHZHZOJONA-N
CHEBI:31399

Spécifications

Cilnidipine
98.5% min. (HPLC)
5g
492.52
CHEBI:31399
99%
132203-70-4
C27H28N2O7
KJEBULYHNRNJTE-DHZHZOJONA-N
5282138
492.52
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes serious eye damage.

GHS Signal Word: Danger

FDS
Documents

Documents

Promotions

Promotions