missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CID 2745687
Description
CID 2745687 is a competitive, reversible antagonist of the orphan receptor GPR35 (Ki = 12.8 nM). Blocks the GPR35-mediated increase in ERK1/2 phosphorylation and β-arrestin recruitment induced by pamoic acid.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 395.43 |
| Purity | >98% |
| Molecular Weight (g/mol) | 395.429 |
| Chemical Name or Material | CID 2745687 |
Safety and Handling
| Recommended Storage | Store at -20°C |