accessibility menu, dialog, popup

UserName

CH3H3CCH3HNSHNNNNFFOOCH3(E/Z)

CAS RN 264233-05-8

IUPAC Name: methyl 5-({[(tert-butylcarbamothioyl)amino]imino}methyl)-1-(2,4-difluorophenyl)-1H-pyrazole-4-carboxylate
Synonyms: methyl 5-({[(tert-butylcarbamothioyl)amino]imino}methyl)-1-(2,4-difluorophenyl)pyrazole-4-carboxylate
Molecular Weight (g/mol): 395.43
Molecular Formula: C17H19F2N5O2S
InChi Key: CYNLZIBKERMMOA-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=NNC(=S)NC(C)(C)C)N(N=C1)C1=C(F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results