missing translation for 'onlineSavingsMsg'
Learn More

Chromotrope 2R, TCI America™

Catalog No. C054725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C054725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C054725G Supplier TCI America Supplier No. C054725G
Only null left

Chemical Identifiers

CAS 4197-07-3
Molecular Formula C16H10N2Na2O8S2
Molecular Weight (g/mol) 468.36
MDL Number MFCD00003955
InChI Key XTJONEUTTZZRAB-VZRGYXGKSA-L
Synonym chromotrope 2r, acid phloxine gr, disodium 4,5-dihydroxy-3-e-2-phenyldiazen-1-yl naphthalene-2,7-disulfonate
PubChem CID 44135388
IUPAC Name disodium;5-hydroxy-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 468.36
Physical Form Crystalline Powder
Chemical Name or Material Chromotrope 2R
RTECSNumber QJ6418000
TSCA No