missing translation for 'onlineSavingsMsg'
Learn More

Chromium(VI) Oxide, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1002290250
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1002290250 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1002290250 Supplier Technidata Supplier No. 1.00229.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity reagent suitable for analytical applications.

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Physical Form Solid
Quantity 250 g
UN Number UN 1463
Formula Weight 99.99
Percent Purity 99.0%
Odor Odorless
Packaging Glass bottle
Appearance Dark red solid
Chemical Name or Material Chromium(VI) Oxide
Hazard Category H271: May cause fire or explosion; strong oxidiser.
H350: May cause cancer.
H340: May cause genetic defects.
H361f: Suspected of damaging fertility.
H330: Fatal if inhaled.
H301 + H311: Toxic if swallowed or in contact with skin
H372: Causes damage to organs through prolonged or repeated exposure.
H314: Causes severe skin burns and eye damage.
H334: May cause allergy or asthma symptoms or breathing difficulties if inhaled.
H317: May cause an allergic skin reaction.
H335: May cause respiratory irritation.
H410: Very toxic to aquatic life with long lasting effects.