missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium ICP Standard, 1000 ppm Cr6+ in 3% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company PCR61KH500

Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. PCR61KH500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7789-09-5 , 7647-01-0 , 7732-18-5
252.06
24600
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
Cr2H8N2O7
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium

Specifications

Chromium(VI) ICP Standard
7789-09-5 , 7647-01-0 , 7732-18-5
98.0, 3.48, 0.24
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24600
Natural Poly Bottle
94.0, 3.34, 0.23
Cr2H8N2O7
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
252.06
1mL = 1mg Cr (1, 000ppm Cr)(NH4)2Cr2O7 in 3% HCl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents