missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium AA Standard, 1000 ppm Cr6+ in H2O, Ricca Chemical

Catalog No. p-3255643
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ACR61KW100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACR61KW100 Supplier Ricca Chemical Company Supplier No. ACR61KW100
Only null left

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Cr
CAS Min % 98.0, 0.28
CAS Max % 100.0, 0.29
Equivalence 1 mL = 1 mg Cr
Physical Form Liquid
Color Yellow
Boiling Point 100°C
Quantity 100 mL
Chemical Name or Material Chromium(VI) AA Standard