missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium Trioxide ACS AR Crystal, Macron Fine Chemicals™
Specifications
Specifications
| CAS | 1333-82-0 |
| Quantity | 2.5kg |
| Molecular Formula | CrO3 |
| Synonym | chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide |
| InChI Key | WGLPBDUCMAPZCE-UHFFFAOYSA-N |
| SMILES | O=[Cr](=O)=O |
| IUPAC Name | trioxochromium |
| Molecular Weight (g/mol) | 99.993 |
| PubChem CID | 14915 |
| ChEBI | CHEBI:48240 |
| Show More |