missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride hexahydrate, 99.5% min

Catalog No. AA4211436
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4211436 500 g
AA4211422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4211436 Supplier Thermo Scientific Chemicals Supplier No. 04211436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10060-12-5 | Cl3CrH12O6 | 266.44 g/mol

Chromium(III) chloride hexahydrate is used as a catalyst, as a mordant in printing and dyeing industry. It is involved in the preparation of anhydrous tetrahydrofuran complex of chromium(III) chloride by reacting with trimethylsilylchloride. It finds application in the manufacturing of a variety of chromium-containing pigment in Paint industry and other chromium salts in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Melting Point 83°C
Color Black to Green
Physical Form Crystalline
Quantity 500 g
UN Number UN3260
Merck Index 14,2222
Solubility Information Slightly soluble in acetone. Soluble in water,alcohol. Insoluble in ether
IUPAC Name chromium(3+) hexahydrate trichloride
Formula Weight 266.45 (158.35 Anhydrous)
Percent Purity ≤99.5%
Odor Odorless
Sensitivity Hygroscopic
Density 1.76 g/mL
Chemical Name or Material Chromium(III) chloride hexahydrate
Show More Show Less
Hazard Category H290-H302-H317
Hazard Statement GHS H Statement
H334-H314-H318-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-038-3
RTECSNumber GB5450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only