missing translation for 'onlineSavingsMsg'
Learn More

Chromium ICP-MS Standard, 1000 ppm Cr in 3% HNO3, Ricca Chemical

Catalog No. MSCR1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSCR1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSCR1KN100 Supplier Ricca Chemical Company Supplier No. MSCR1KN100

Chemical Identifiers

CAS 7789-02-8 , 7697-37-2 , 7732-18-5
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Cr
CAS Min % 93.0, 4.2, 0.72
CAS Max % 97.0, 4.37, 0.75
Equivalence 1 mL = 1 mg Cr
Physical Form Liquid
Color Blue
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Chromium ICP-MS Standard
Show More Show Less