missing translation for 'onlineSavingsMsg'
Learn More

Chromium hexacarbonyl, 99+%

Catalog No. AC221050100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221050100 10 g Glass bottle
AC221050500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221050100 Supplier Thermo Scientific Chemicals Supplier No. 221050100
Only null left

CAS: 13007-92-6 | C6CrO6 | 220.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13007-92-6
Molecular Formula C6CrO6
Molecular Weight (g/mol) 220.06
MDL Number MFCD00010945
InChI Key KOTQLLUQLXWWDK-UHFFFAOYSA-N
Synonym chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10g
PubChem CID 25589
ChEBI CHEBI:33031
IUPAC Name carbon monoxide;chromium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]

Specifications

Boiling Point 220°C
Melting Point 150°C
Color White through Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 10 g
Assay Percent Range 99+%
Linear Formula Cr(CO)6
Merck Index 15, 2236
Solubility Information Solubility in water: insoluble. Other solubilities: sparingly soluble in organic solvents
Formula Weight 220.06
Percent Purity ≥99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Chromium hexacarbonyl
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause cancer by inhalation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water
EINECSNumber 235-852-4

RUO – Research Use Only