Learn More
MilliporeSigma™ Supelco™ ChromegaChiral™ Chiral CCO F2 HPLC Column, 10 μm Particle Size
ChromegaChiral™ CCO F2 is a 2-Fluoro 5-methylphenyl cellulose phase which can be used in SFC or HPLC.
Supplier:  MilliporeSigma™ Supelco™ ES5571
 View more versions of this product
Catalog No. 111005548
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The addition of a fluorine atom into a phenyl carbamate cellulose structure can be useful in promoting a fluorophilic retention mechanism which can provide improved retention for fluorinated compounds. A fluorophilic retention mechanism can be particularly useful in medicinal chemistry and drug discovery, where more than a third of newly approved small molecule drugs contain fluorine.
Specifications
Specifications
20 mm
10 μm
2-fluoro 5-methylphenyl cellulose
2-fluoro 5-methylphenyl cellulose Phase
Chiral HPLC
25 cm
1000 Å
ChromegaChiral
Chiral
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
ChromegaChiral is a trademark of ES Industries