Learn More
MilliporeSigma™ Supelco™ ChromegaChiral™ Chiral CCC HPLC Column, 5 μm Particle Size
ChromegaChiral™ CCC is a modified cellulose including the combination of 3-chloro-4 methylphenylcarbamate and 3,5-dichloro- phenylcarbamate bonding groups coated on high purity, high performance spherical silica particles.
Supplier: MilliporeSigma™ Supelco™ ES5461
Description
ChromegaChiral™ CCC's combination of bonded groups stabilizes the solubility of coated phase making for a durable phase similar to other widely used coated phases. The use of cellulose modified with chlorinated phenyl groups provides for the separation for many previously unresolved/poorly resolved chiral mixtures by providing unique separation characteristics.Specifications
| 20 mm | |
| 5 μm | |
| 3 chloro-4 methylphenylcarbamate and 3,5-dichlorophenylcarbamate | |
| 3 chloro-4 methylphenylcarbamate and 3,5-dichlorophenylcarbamate Phase | |
| Chiral HPLC |
| 25 cm | |
| 1000 Å | |
| ChromegaChiral | |
| Chiral |
ChromegaChiral is a trademark of ES Industries