missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromate Standard, 1000 ppm CrO42-, Ricca Chemical
Chemical Identifiers
| CAS | 7789-00-6 , 7732-18-5 |
|---|---|
| Molecular Formula | CrK2O4 |
| Molecular Weight (g/mol) | 194.19 |
| MDL Number | MFCD00011368 |
| InChI Key | XMXNVYPJWBTAHN-UHFFFAOYSA-N |
| PubChem CID | 24597 |
| ChEBI | CHEBI:75249 |
| IUPAC Name | dipotassium dioxochromiumbis(olate) |
| SMILES | [K+].[K+].[O-][Cr]([O-])(=O)=O |
| Catalog Number | Mfr. No. | dimQuantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | dimQuantity | Packaging | Price | Quantity & Availability | ||||
|
20704
|
N/A | Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
207016
|
N/A | Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7789-00-6 , 7732-18-5 | |
| 194.19 | |
| XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
| CHEBI:75249 | |
| [K+].[K+].[O-][Cr]([O-])(=O)=O |
| CrK2O4 | |
| MFCD00011368 | |
| 24597 | |
| dipotassium dioxochromiumbis(olate) |
Specifications
| 7789-00-6 , 7732-18-5 | |
| 100.0, 0.17 | |
| CrK2O4 | |
| XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
| dipotassium dioxochromiumbis(olate) | |
| 24597 | |
| Chromate Standard |
| 98.0, 0.17 | |
| 1 mL = 1 mg CrO | |
| MFCD00011368 | |
| [K+].[K+].[O-][Cr]([O-])(=O)=O | |
| 194.19 | |
| CHEBI:75249 |