missing translation for 'onlineSavingsMsg'
Learn More

Choline Bitartrate 98.0+%, TCI America™

Catalog No. C055325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C055325G 25 g
C0553500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C055325G Supplier TCI America Supplier No. C055325G

Chemical Identifiers

CAS 87-67-2
Molecular Formula C9H19NO7
Molecular Weight (g/mol) 253.251
MDL Number MFCD00036332
InChI Key QWJSAWXRUVVRLH-UHFFFAOYSA-M
Synonym choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
PubChem CID 10198924
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
SMILES C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O

Specifications

Melting Point 150°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 253.25
Percent Purity ≥98.0% (T)
Chemical Name or Material Choline Bitartrate