missing translation for 'onlineSavingsMsg'
Learn More

Choline (47-50% in Water), TCI America™

Catalog No. p-7124356
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C032625G 25 g
C0326500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C032625G Supplier TCI America Supplier No. C032625G

Chemical Identifiers

CAS 123-41-1
Molecular Formula C5H15NO2
Molecular Weight (g/mol) 121.18
MDL Number MFCD00002831
InChI Key KIZQNNOULOCVDM-UHFFFAOYSA-M
Synonym choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide
PubChem CID 31255
IUPAC Name (2-hydroxyethyl)trimethylazanium hydroxide
SMILES [OH-].C[N+](C)(C)CCO

Specifications

Quantity 25 g
UN Number 3267
Formula Weight 121.18
Physical Form Liquid
Chemical Name or Material Choline (47-50% in Water)
RTECSNumber GA4025500
TSCA Yes
Recommended Storage Refrigerator