missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol Benzoate 96.0+%, TCI America™

Catalog No. C032025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C032025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C032025G Supplier TCI America Supplier No. C032025G

Chemical Identifiers

CAS 604-32-0
Molecular Formula C34H50O2
Molecular Weight (g/mol) 490.77
MDL Number MFCD00003635
InChI Key UVZUFUGNHDDLRQ-UHFFFAOYNA-N
Synonym cholesteryl benzoate, cholesterol benzoate, 3b-benzoyloxy cholest-5-ene, unii-n09h13shlb, n09h13shlb, cholest-5-ene-3-beta-yl benzoate, cholest-5-en-3-ol 3.beta.-, benzoate, 5-cholesten-3.beta.-ol benzoate, cholest-5-en-3-ol 3beta-, benzoate
PubChem CID 2723613
IUPAC Name 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl benzoate
SMILES CC(C)CCCC(C)C1CCC2C3CC=C4CC(CCC4(C)C3CCC12C)OC(=O)C1=CC=CC=C1

Specifications

Melting Point 150°C
Color White
Quantity 25 g
Formula Weight 490.77
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cholesterol Benzoate
EINECSNumber (4)-0455
TSCA Yes