missing translation for 'onlineSavingsMsg'
Learn More

Chlorzoxazone 98.0+%, TCI America™

Catalog No. C223725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C223725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C223725G Supplier TCI America Supplier No. C223725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-25-0
Molecular Formula C7H4ClNO2
Molecular Weight (g/mol) 169.564
MDL Number MFCD00005717
InChI Key TZFWDZFKRBELIQ-UHFFFAOYSA-N
Synonym chlorzoxazone, chlorzoxazon, paraflex, chloroxazone, 5-chloro-2-benzoxazolone, biomioran, escoflex, myoflexin, myoflexine, pathorysin
PubChem CID 2733
ChEBI CHEBI:3655
IUPAC Name 5-chloro-3H-1,3-benzoxazol-2-one
SMILES C1=CC2=C(C=C1Cl)NC(=O)O2

Specifications

Melting Point 191°C
Quantity 25 g
Formula Weight 169.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorzoxazone
RTECSNumber DM5250000
TSCA Yes