Learn More
Thermo Scientific Chemicals Chlorpropham
$55.70 - $55.70
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465580050
|
Thermo Scientific Chemicals
465580050 |
5 g |
Each for $55.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 101-21-3 | |
| C10H12ClNO2 |
Specifications
| 101-21-3 | |
| Solubility aq. soln.: insoluble. Other solubilities: miscible with acetone, methanol, ethanol | |
| C10H12ClNO2 | |
| CWJSHJJYOPWUGX-UHFFFAOYSA-N | |
| propan-2-yl N-(3-chlorophenyl)carbamate | |
| Chlorpropham |
| 5 g | |
| 36.0°C to 38.0°C | |
| 213.66 | |
| CC(C)OC(=O)NC1=CC=CC(Cl)=C1 | |
| 213.66 |
Safety and Handling
GHS H Statement Harmful if swallowed.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 202-925-7
RUO – Research Use Only