missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorpropham

Catalog No. AC465580050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465580050 5 g
AC465580250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465580050 Supplier Thermo Scientific Chemicals Supplier No. 465580050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101-21-3
Molecular Formula C10H12ClNO2

Specifications

Quantity 5 g
Solubility Solubility aq. soln.: insoluble. Other solubilities: miscible with acetone, methanol, ethanol
Melting Point 36.0°C to 38.0°C
Formula Weight 213.66
InChI Key CWJSHJJYOPWUGX-UHFFFAOYSA-N
SMILES CC(C)OC(=O)NC1=CC=CC(Cl)=C1
IUPAC Name propan-2-yl N-(3-chlorophenyl)carbamate
Molecular Weight (g/mol) 213.66
Chemical Name or Material Chlorpropham
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Suspected of causing cancer.

May cause damage to organs through prolonged or repeated exposure.

Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 202-925-7

RUO – Research Use Only