missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotripropylsilane, 98%
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | chlorotripropylsilane |
| Couleur | Incolore |
| Densité | 0.8820g/mL |
| Point d’ébullition | 199.0°C to 200.0°C |
| Point d’éclair | 76°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 97.5% min. (GC) |
| Emballage | Bouteille en verre |
| Indice de réfraction | 1.3990 to 1.4410 (20°C, 589nm) |
| Quantité | 5 g |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes severe skin burns and eye damage. Contact with water liberates toxic gas. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. Store in a dry place. Store in a closed container. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Protect from moisture. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 213-624-5 |
RUO – Usage réservé à la recherche