missing translation for 'onlineSavingsMsg'
Learn More

Chloroquine phosphate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014370
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014370 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014370 Supplier Merck Emd Millipore Supplier No. PHR12581G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 50-63-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C18H26 ClN3 · 2H3 PO4
MDL Number MFCD00069852
Melting Point 200°C
Synonym Chloroquine diphosphate salt; N4-(7-Chloro-4-quinolinyl)-N1,N1-dimethyl-1,4-pentanediamine diphosphate salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 515.86
Grade Certified Reference Material
Show More Show Less