missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorophenol Red Indicator, 0.04% (w/v), Ricca Chemical
$77.83 - $134.43
Specifications
| Chemical Name or Material | Chlorophenol Red Indicator |
|---|---|
| CAS | 123333-64-2 , 7732-18-5 |
| CAS Min % | 98.0,0.04 |
| CAS Max % | 100.0,0.04 |
| Concentration | 0.04% (w/v) |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
2000-4
|
Ricca Chemical Company
RI20004 |
120 mL | Natural Poly Bottle |
Each for $77.83
|
|
||||
|
2000-16
|
Ricca Chemical Company
200016 |
500 mL | Natural Poly Bottle |
Each for $103.27
|
|
||||
|
200032
|
Ricca Chemical Company
200032 |
1 L | Natural Poly Bottle |
Each for $134.43
|
|
||||
Specifications
| Chlorophenol Red Indicator | |
| 98.0,0.04 | |
| 0.04% (w/v) | |
| Indicator | |
| Aqueous Solution | |
| 6 |
| 123333-64-2 , 7732-18-5 | |
| 100.0,0.04 | |
| 0.04% W/V | |
| Natural Poly Bottle | |
| Red | |
| Liquid |