missing translation for 'onlineSavingsMsg'
Learn More

Chlorophenol Red Indicator, 0.04% (w/v), Ricca Chemical

Catalog No. 20004
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
20004 120 mL Natural Poly Bottle
200016 500 mL Natural Poly Bottle
200032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20004 Supplier Ricca Chemical Company Supplier No. RI20004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Chlorophenol Red Indicator
CAS 123333-64-2, 7732-18-5
CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Concentration 0.04% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Red
pH 6
Quantity 120 mL
Physical Form Liquid
Percent Purity 0.04% W/V
Grade Indicator
Show More Show Less