missing translation for 'onlineSavingsMsg'
Learn More

Chlorophenol Red Indicator, 0.04% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 200016

 View more versions of this product

Catalog No. 2000-16


Only null left

Description

Description

Specifications

Specifications

Chlorophenol Red Indicator
98.0,0.04
0.04% (w/v)
Indicator
Aqueous Solution
6
Liquid
123333-64-2 , 7732-18-5
100.0,0.04
0.04% W/V
Natural Poly Bottle
Red
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions